C91352
1-(2-Cyanoethyl)pyrrole
≥99%
Synonym(s):
1-Pyrrolepropionitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8N2
CAS Number:
Molecular Weight:
120.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99%
form
liquid
refractive index
n20/D 1.51 (lit.)
bp
132-133 °C/10 mmHg (lit.)
density
1.048 g/mL at 25 °C (lit.)
SMILES string
N#CCCn1cccc1
InChI
1S/C7H8N2/c8-4-3-7-9-5-1-2-6-9/h1-2,5-6H,3,7H2
InChI key
IYOLJLGYJMJLSU-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Muamer Dervisevic et al.
Journal of food and drug analysis, 25(3), 510-519 (2017-09-16)
The aim of this study was the electrochemical detection of the adenosine-3-phosphate degradation product, xanthine, using a new xanthine biosensor based on a hybrid bio-nanocomposite platform which has been successfully employed in the evaluation of meat freshness. In the design
Elif Burcu Aydın
Talanta, 215, 120909-120909 (2020-04-22)
A novel impedimetric immunosensor based on a conjugated polypyrrole polymer containing epoxy active side groups (PPCE) modified indium tin oxide (ITO) electrode was developed for detection of interleukin 6 (IL 6), a prostate cancer biomarker. IL 6 receptor was used
Elif Burcu Aydın et al.
Talanta, 222, 121596-121596 (2020-11-11)
An ultrasensitive immunosensor based on acetylene black (AB)/epoxy-substituted-poly(pyrrole) polymer (EpxS-PPyr) composite coated disposable indium tin oxide (ITO) electrode was fabricated for interleukin 6 (IL 6) detection. The EpxS-PPyr polymer was a promising matrix material to increase the loading capacity of
Jae Y Lee et al.
Journal of biomedical materials research. Part A, 103(6), 2126-2132 (2014-10-09)
Electrically conducting polymers (CPs) have been recognized as novel biomaterials that can electrically communicate with biological systems. For their tissue engineering applications, CPs have been modified to promote cell adhesion for improved interactions between biomaterials and cells/tissues. Conventional approaches to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service