Skip to Content
Merck
All Photos(2)

Documents

C11006

Sigma-Aldrich

Carbohydrazide

98%

Synonym(s):

1,3-Diaminourea, Carbodihydrazide, Carbonohydrazide, N,N′-Diaminourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO(NHNH2)2
CAS Number:
Molecular Weight:
90.08
Beilstein:
1747069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

crystals

mp

150-153 °C (lit.)

SMILES string

NNC(=O)NN

InChI

1S/CH6N4O/c2-4-1(6)5-3/h2-3H2,(H2,4,5,6)

InChI key

XEVRDFDBXJMZFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbohydrazide can be used:
  • As an oxygen scavenger.
  • In the synthesis of polydentate Schiff base ligands with various aldehydes and ligands by condensation.
  • In the synthesis of trifluoromethyl-containing (E)-N′-arylidene-1H-pyrazole-1-carbohydrazides by cyclocondensation reaction which shows antioxidant and antimicrobial properties.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro K Jordão et al.
Bioorganic & medicinal chemistry, 19(18), 5605-5611 (2011-08-16)
Tuberculosis treatment remains a challenge that requires new antitubercular agents due to the emergence of multidrug-resistant Mycobacterium strains. This paper describes the synthesis, the antitubercular activity and the theoretical analysis of N-substituted-phenylamino-5-methyl-1H-1,2,3-triazole-4-carbohydrazides (8a-b, 8e-f, 8i-j and 8n-o) and new analogues
New trifluoromethyl-containing (E)-N?-arylidene-[3-alkyl (aryl/heteroaryl)-4, 5-dihydro-1H-pyrazol-1-yl] carbohydrazides: Synthesis, crystal structure and antimicrobial/antioxidant activity.
Bonacorso HG, et al.
Journal of Fluorine Chemistry, 135(10), 303-314 (2012)
Paolo Mombelli et al.
ChemMedChem, 7(1), 151-158 (2011-11-19)
A series of inhibitors of plant enzymes of the non-mevalonate pathway from herbicide research efforts at BASF were screened for antimalarial activity in a cell-based assay. A 1,3-diiminoisoindoline carbohydrazide was found to inhibit the growth of Plasmodium falciparum with an
Marta Kisiel et al.
Journal of controlled release : official journal of the Controlled Release Society, 162(3), 646-653 (2012-08-21)
Understanding the influence of formulation and storage conditions on rhBMP-2 bioactivity is extremely important for its clinical application. Reports in the literature show that different research groups employ different parameters such as formulation conditions, storage, doses for in vivo applications
Ameen Ali Abu-Hashem et al.
Acta pharmaceutica (Zagreb, Croatia), 60(3), 311-323 (2010-12-08)
2-Amino-5-acetyl-4-methyl-thiophene-3-carboxylic acid ethyl ester (1) and 5-acetyl-2-amino-4-methylthiophene-3-carbohydrazide (2) were synthesized and used as starting materials for the synthesis of new series of 1-(5-amino-4-(3,5-dimethyl-1H-pyrazole-1-carbonyl)-3-methylthiophen-2-yl) ethanone (3a), 1-(5-amino-4-(4-chloro-3,5-dimethyl-1H-pyrazole-1-carbonyl)-3-methylthiophen-2-yl) ethanone (3b), 1-(4-methyl-2-amino-5-acetylthiophene-3-carbonyl)pyrazolidine-3,5-dione (4), (Z)-N'-(4-methyl-2-amino-5-acetylthiophene-3-carbonyl) formohydrazonic acid (5a), (Z)-ethyl-N'-4-methyl-2-amino-5-acetylthiophene-3-carbonylformo hydrazonate (5b), 6-acetyl-3-amino-2,5-dimethylthieno[2,3-d]pyrimidin-4(3H)-one (8), 5-methyl-3-amino-2-mercapto-6-acetylthieno

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service