Merck
BR144635
BRAND® BISTABIL® ground glass stopper, conical joint
borosilicate glass 3.3, joint: ST/NS 29/32, (hollow, hexagonal grip)
Synonym(s):
glass closure, glass plug, sealing plug, stopper cap
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.43
UNSPSC Code:
41121800
Material:
borosilicate glass 3.3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
borosilicate glass 3.3
joint
joint: ST/NS29/32
manufacturer/tradename
BRAND 144635
General description
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)