All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C32H43ClN2O2Pd
CAS Number:
Molecular Weight:
629.57
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
Recommended Products
form
powder
Quality Level
reaction suitability
reagent type: catalyst
reaction type: Cross Couplings
mp
>300 °C
Application
Pd(IPr)(acac)Cl is a Pd-catalyst that can be used in a broad range of reactions, including cross-coupling, C-H functionalization, electrophilic additions.
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Pd(NHC)(acac)Cl]: Well-Defined, Air-Stable, and Readily Available Precatalysts for Suzuki and Buchwald−Hartwig Cross-coupling (Transamidation) of Amides and Esters by N−C/O−C Activation.
Zhou T, et al.
Organic Letters, 21, 3304-3309 (2019)
Nicolas Marion et al.
The Journal of organic chemistry, 71(10), 3816-3821 (2006-05-06)
A very straightforward synthesis of (IPr)Pd(acac)Cl from two commercially available starting materials, Pd(acac)2 and IPr.HCl [acac = acetylacetonate; IPr = N,N'-bis(2,6-diisopropylphenyl)imidazol-2-ylidene], has been developed. The resulting complex, (IPr)Pd(acac)Cl (1), has proven to be a highly active PdII precatalyst in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service