Skip to Content
Merck
All Photos(1)

Key Documents

922803

Sigma-Aldrich

[Pt(IPr*)(DMS)Cl2]

Synonym(s):

Platinum N,N′-bis(2,6-bis(diphenylmethyl)-4-methylphenyl)imidazolium dimethylsulfide dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C71H62Cl2N2PtS
Molecular Weight:
1241.31
UNSPSC Code:
12141733
NACRES:
NA.22

form

powder

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

mp

248-255 °C (decomposition)

Application

[Pt(IPr*)(DMS)Cl2] is a platinum NHC catalsyst used for olefin hydrosilylation.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benon P Maliszewski et al.
Dalton transactions (Cambridge, England : 2003), 49(41), 14673-14679 (2020-10-17)
A general, user-friendly synthetic route to [Pt(NHC)(L)Cl2] and [Pt(NHC)(dvtms)] (L = DMS, Py; DMS = dimethyl sulfide, dvtms = divinyltetramethylsiloxane, Py = pyridine) complexes has been developed. The procedure is applicable to a wide range of ligands and enables facile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service