83060
tert-Butyl 12-amino-4,7,10-trioxadodecanoate
technical, ≥80% (T)
Synonym(s):
Amino-PEG3-tert-butyl ester, tert-Butyl 3-[2-(2-(2-aminoethoxy)ethoxy)ethoxy]propionate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H27NO5
CAS Number:
Molecular Weight:
277.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
grade
technical
Quality Level
Assay
≥80% (T)
form
solid
reaction suitability
reagent type: cross-linking reagent
storage temp.
−20°C
SMILES string
CC(C)(C)OC(=O)CCOCCOCCOCCN
InChI
1S/C13H27NO5/c1-13(2,3)19-12(15)4-6-16-8-10-18-11-9-17-7-5-14/h4-11,14H2,1-3H3
InChI key
CWFSAZJIJBTKRC-UHFFFAOYSA-N
Application
tert-Butyl 12-amino-4,7,10-trioxadodecanoate is generally used as a spacer or linker in bioconjugate chemistry. Some of its applications are:
- Synthesis of the tetanus-toxin conjugate of MUC1 glycopeptide antigen, as a potential antitumor vaccine.
- Synthesis of self-assembled monolayer-based surface-enhanced raman scattering (SERS) labels for immuno-SERS microscopy.
- Synthesis of polycationic adamantane-based dendrons for cell imaging.
- Synthesis of phthalocyanine (Pc)-peptide conjugates as potential fluorescence imaging agents for cancers overexpressing epidermal growth factor receptors (EGFR).
Other Notes
Intermediate in the synthesis of a spacer for combinatorial chemistry
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
337.5 °F
Flash Point(C)
169.7 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Towards the Development of Antitumor Vaccines: A Synthetic Conjugate of a Tumor?Associated MUC1 Glycopeptide Antigen and a Tetanus Toxin Epitope.
Keil S, et al.
Angewandte Chemie (International Edition in English), 40(2), 366-369 (2001)
Phthalocyanine?peptide conjugates for epidermal growth factor receptor targeting.
Ongarora B G, et al.
Journal of Medicinal Chemistry, 55(8), 3725-3738 (2012)
Water soluble SERS labels comprising a SAM with dual spacers for controlled bioconjugation.
Jehn C, et al.
Physical Chemistry Chemical Physics, 11(34), 7499-7504 (2009)
Angewandte Chemie (International Edition in English), 113, 379-379 (2001)
Polycationic adamantane-based dendrons of different generations display high cellular uptake without triggering cytotoxicity.
Grillaud M, et al.
Journal of the American Chemical Society, 136(2), 810-819 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service