Skip to Content
Merck
All Photos(1)

Key Documents

791814

Sigma-Aldrich

KetoABNO

95%

Synonym(s):

9-Azabicyclo[3,3,1]nonan-3-one-9-oxyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12NO2
CAS Number:
Molecular Weight:
154.19
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

reaction suitability

reagent type: oxidant

mp

109-114 °C

storage temp.

2-8°C

SMILES string

O=C1C[C@@H](N2[O])CCC[C@@H]2C1

InChI

1S/C8H12NO2/c10-8-4-6-2-1-3-7(5-8)9(6)11/h6-7H,1-5H2

InChI key

AMZBXNVXJGUYMF-UHFFFAOYSA-N

Application

Oxidant has been demonstrated in the application of aerobic oxidation of alcholols and amines at room temperature and ambient pressure. This product is also a convenient bench top stable solid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonobe T, et al.
Chemical Science, 3, 3249-3249 (2012)
Lauber MB, and Stahl SS
ACS Catalysis, 3, 2612-2616 (2013)

Related Content

he Stahl Lab focuses on the development of catalysts and catalytic reactions for selective oxidation of organic molecules, with particular emphasis on aerobic oxidation reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service