77386
2-Bromo-1-ethyl-pyridinium tetrafluoroborate
≥97.0% (T)
Synonym(s):
BEP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥97.0% (T)
form
crystals
reaction suitability
reaction type: Coupling Reactions
mp
103-107 °C
application(s)
peptide synthesis
SMILES string
F[B-](F)(F)F.CC[n+]1ccccc1Br
InChI
1S/C7H9BrN.BF4/c1-2-9-6-4-3-5-7(9)8;2-1(3,4)5/h3-6H,2H2,1H3;/q+1;-1
InChI key
YJDXVQLBIAJTHP-UHFFFAOYSA-N
General description
2-Bromo-1-ethyl-pyridinium tetrafluoroborate is a coupling reagent employed in the synthesis of amides and esters through amidation and esterification reactions, respectively. It is generally prepared by the reaction of triethyloxonium tetrafluoroborate with 2?bromo pyridine.
Application
2-Bromo-1-ethyl-pyridinium tetrafluoroborate can be used as a coupling reagent for:
- The synthesis of N-methylated peptides in solution and solid phase.
- The synthesis of cyclosporin A fragment and dolastatin 15 pentapeptide moiety.
Other Notes
Coupling reagent for peptide synthesis; less racemization and faster reaction than with other reagents;e.g. BOP, PyBrOP, PyClU, BTFFH, CMBI
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
AIDS (London, England), 33(6), 993-1000 (2019-04-05)
Epstein-Barr virus (EBV) has been implicated in lymphomagenesis of HIV-related classical Hodgkin lymphoma (HIV-cHL). The utility of EBV molecular and serological biomarkers has scarcely been examined in HIV-cHL in the recent combined antiretroviral therapy (cART) era. We evaluated EBV DNA
Journal of the Chinese Medical Association : JCMA, 82(6), 510-514 (2019-06-11)
Congenital TORCH (toxoplasmosis, other viruses [varicella-zoster virus, VZV, etc.], rubella, cytomegalovirus [CMV], Herpes simplex virus [HSV]) infections are major causes of prenatal, perinatal, and postnatal morbidity and mortality. Although treatment or prevention strategies are available for these pathogens, all drugs
Nature communications, 8(1), 2146-2146 (2017-12-17)
Hepatitis B virus (HBV) is a major global health concern, and the development of curative therapeutics is urgently needed. Such efforts are impeded by the lack of a physiologically relevant, pre-clinical animal model of HBV infection. Here, we report that
SLAS technology, 23(2), 188-197 (2018-01-19)
The lack of (inter-)laboratory standardization has hampered the application of universal cutoff values for Alzheimer's disease (AD) cerebrospinal fluid (CSF) biomarkers and their transfer to general clinical practice. The automation of the AD biomarker immunoassays is suggested to generate more
Journal of medical virology, 89(2), 324-331 (2016-07-23)
Cytomegalovirus (CMV) is the most common congenital viral infection. Mother-to-child transmission can cause severe child disability. Intact CMV-specific cell-mediated immunity (CMI) was shown to prevent uncontrolled replication in healthy individuals. This study aimed to determine whether CMV-specific CMI is impaired
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service