Skip to Content
Merck
All Photos(3)

Key Documents

771147

Sigma-Aldrich

1-Trifluoromethyl-1,2-benziodoxol-3-(1H)-one

60 wt. %, contains 40 wt. % Celatom® FW-80 as additive

Synonym(s):

Togni Reagent II

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

contains

40 wt. % Celatom® FW-80 as additive

reaction suitability

reaction type: C-C Bond Formation

concentration

60 wt. %

mp

150-158 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)[I]1OC(=O)c2ccccc12

InChI

1S/C8H4F3IO2/c9-8(10,11)12-6-4-2-1-3-5(6)7(13)14-12/h1-4H

InChI key

XHEOXSQMBWJOKP-UHFFFAOYSA-N

Application

Electrophilic trifluoromethylating reagent has shown to be one of the more robust trifluoromethylation reagents on the market. However, a recent report has showed this product to have potential dangerous self-reactivity neat. We now offer a less self-reactive material mixed down in Celatom®, a silica-based adsorbent, which significantly reduces self-reactivity allowing for safer shipping and storage.

Legal Information

Celatom is a registered trademark of EP Minerals, LLC

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Fiederling, N.;
Organic Process Research & Development, 17, 318-319 (2013)

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Related Content

Research in the Togni group focuses on the development of new ligands and reagents. These two general directions thus impact the ability to construct molecules in more efficient or unprecedented ways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service