Skip to Content
Merck
All Photos(1)

Key Documents

745715

Sigma-Aldrich

Oxetane-3-methanol

97%

Synonym(s):

3-(Hydroxymethyl)oxetane, 3-Oxetanemethanol, 3-Oxetanylmethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.450

density

1.092 g/mL at 25 °C

storage temp.

−20°C

SMILES string

OCC1COC1

InChI

1S/C4H8O2/c5-1-4-2-6-3-4/h4-5H,1-3H2

InChI key

SWYHWLFHDVMLHO-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

223.9 °F

Flash Point(C)

106.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service