Skip to Content
Merck
All Photos(3)

Documents

702471

Sigma-Aldrich

PTAA

greener alternative

a poly(triaryl amine) semiconductor

Synonym(s):

Poly(triaryl amine), Poly[bis(4-phenyl)(2,4,6-trimethylphenyl)amine]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H4N(C6H2(CH3)3)C6H4]n
CAS Number:
UNSPSC Code:
32111503
NACRES:
NA.23

form

solid

mol wt

average Mn 7,000-10,000 (GPC)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

>400 °C
>400 °C

Mw/Mn

2‑2.2

application(s)

battery manufacturing
semiconductor

greener alternative category

semiconductor properties

P-type (mobility=10−3 - 10−2 cm2/V·s)

General description

PTAA, poly(triaryl amine), semiconductor is an organic p-type semiconductor with hole mobilities of 10−3 up to 10−2 cm2 V−1 s−1 which results in a high carrier mobility. It is a stable glassy polymer and has good ionization potential for thick film diodes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Hole transport organic materials allow perfect energy level alignment with the absorber layer and therefore efficient charge collection, are prone to degradation in ambient conditions.Click here for more information.

Application

PTAA can be coated as a substrate material which is used for the transportation of hole in the fabrication of many devices like perovskite solar cells, polymeric light emitting diodes and organic field effect transistors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triarylamine-containing poly (perfluorocyclobutane) as hole-transporting material for polymer light-emitting diodes.
Liu S, et al.
Macromolecules, 33(10), 3514-3517 (2000)
Dielectric function and degradation process of poly (triarylamine)(PTAA).
Sendner M, et al.
Organic Electronics, 15(11), 2959-2963 (2014)
Solvent engineering for high-performance inorganic-organic hybrid perovskite solar cells.
Jeon NJ, et al.
Nature Materials, 13(9), 897-897 (2014)
M.L. Chabinyc, L.H. Jimison, J. Rivnay, A. Salleo
Mrs Bulletin, 33, 683-683 (2008)
Enhanced charge separation in ternary P3HT/PCBM/CuInS2 nanocrystals hybrid solar cells.
Lefrancois A, et al.
Scientific Reports, 5(9), 7768-7768 (2015)

Articles

Polytriarylamine Semiconductors

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service