689416
Pyrrole-3-carboxylic acid
≥96%
Synonym(s):
3-Carboxypyrrole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H5NO2
CAS Number:
Molecular Weight:
111.10
Beilstein:
108642
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥96%
≥96.5% (GC)
96.0-104.0% (titration)
form
powder
functional group
carboxylic acid
SMILES string
OC(=O)c1cc[nH]c1
InChI
1S/C5H5NO2/c7-5(8)4-1-2-6-3-4/h1-3,6H,(H,7,8)
InChI key
DOYOPBSXEIZLRE-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Eleni Sapountzi et al.
Frontiers in chemistry, 8, 678-678 (2020-08-28)
In this work, polyacrylonitrile (PAN) nanofiber mats coated with conductive polypyrrole layers were produced at the surface of gold electrodes by a two-step approach combining electrospinning and vapor phase polymerization. In the first step, smooth and uniform PAN fibers exhibiting
Jungkyun Oh et al.
Polymers, 12(6) (2020-07-02)
Despite being highly flammable at lower concentrations and causing suffocation at higher concentrations, hydrogen gas continues to play an important role in various industrial processes. Therefore, an appropriate monitoring system is crucial for processes that use hydrogen. In this study
E Sheikhzadeh et al.
Biosensors & bioelectronics, 80, 194-200 (2016-02-03)
The Gram-negative bacterium, Salmonella Typhimurium (S. Typhimurium) is a food borne pathogen responsible for numerous hospitalisations and deaths all over the world. Conventional detection methods for pathogens are time consuming and labour-intensive. Hence, there is considerable interest in faster and
Ali Özcan et al.
Talanta, 165, 489-495 (2017-02-06)
In this study, preparation of a single-use electrochemical sensor for the selective and sensitive determination of dopamine (DOP) was investigated by electrochemical polymerization of pyrrole-3-carboxylic acid on electrochemically over-oxidized pencil graphite electrode (p(P3CA)/EOPGE). Cyclic voltammetry measurements of Fe(CN)
H Zejli et al.
Talanta, 185, 513-519 (2018-05-16)
This work demonstrates the development of electrochemical aptasensor using ochratoxin A (OTA) aptamers. Different aptamer coupling strategies were tested using polythiophene-carboxylic acid (PT3C) and polypyrrole-3-carboxylic acid (PP3C). The best sensitivity was recorded by polythiophene-3-carboxylic acid (PT3C) on screen-printed carbon electrode
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service