Skip to Content
Merck
All Photos(1)

Key Documents

688762

Sigma-Aldrich

1,3-Diethoxyimidazolium bis(trifluoromethylsulfonyl)imide

98%

Synonym(s):

(OEt)2Im-NTF2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13F6N3O6S2
CAS Number:
Molecular Weight:
437.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98% (13C-NMR)
≥98% (19F-NMR)
≥98% (31P-NMR)
98%

form

liquid

impurities

0.3% Pf6 (19F-NMR)

refractive index

n20/D 1.4249

suitability

complies for IR spectroscopy

SMILES string

CCOn1cc[n+](OCC)c1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C7H13N2O2.C2F6NO4S2/c1-3-10-8-5-6-9(7-8)11-4-2;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-7H,3-4H2,1-2H3;/q+1;-1

InChI key

CJJJLUVYFXOOSP-UHFFFAOYSA-N

Application

Ionic liquid and NHC precatalyst

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N,N-Di(alkoxy)imidazolium salts: new patent-free ionic liquids and NHC-precatalysts
Laus, G., et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 62, 295-308 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service