Skip to Content
Merck
All Photos(1)

Key Documents

609889

Sigma-Aldrich

Methanol-18O

95 atom % 18O

Synonym(s):

Methyl alcohol-18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH318OH
CAS Number:
Molecular Weight:
34.04
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

95 atom % 18O

Quality Level

Assay

99% (CP)

bp

64.7 °C (lit.)

density

0.838 g/mL at 25 °C

mass shift

M+2

SMILES string

C[18OH]

InChI

1S/CH4O/c1-2/h2H,1H3/i2+2

InChI key

OKKJLVBELUTLKV-HQMMCQRPSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service