Skip to Content
Merck
All Photos(1)

Key Documents

606316

Sigma-Aldrich

Ethanolamine-2-13C

99 atom % 13C

Synonym(s):

2-Aminoethanol-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH213CH2NH2
CAS Number:
Molecular Weight:
62.08
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

170 °C (lit.)

mp

11 °C (lit.)

density

1.030 g/mL at 25 °C

mass shift

M+1

SMILES string

N[13CH2]CO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2/i1+1

InChI key

HZAXFHJVJLSVMW-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service