Skip to Content
Merck
All Photos(2)

Key Documents

559938

Sigma-Aldrich

4-Mercaptophenol

97%

Synonym(s):

4-Hydroxythiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4OH
CAS Number:
Molecular Weight:
126.18
Beilstein:
2039306
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

149-150 °C/25 mmHg (lit.)

mp

33-35 °C (lit.)

SMILES string

Oc1ccc(S)cc1

InChI

1S/C6H6OS/c7-5-1-3-6(8)4-2-5/h1-4,7-8H

InChI key

BXAVKNRWVKUTLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Mercaptophenol (MPH) has been used to study the adsorption of MPH on silver coated polystyrene nanospheres by time-dependent surface-enhanced Raman scattering (SERS) spectroscopy.
4-Mercaptophenol may be used in the preparation of silylated monomer, which was employed for the synthesis of hyperbranched poly(ester-imide). 4-Mercaptophenol (H-4MP) reacts with metal tert-butoxides ([M(OBut)4]) to yield the following Group 4 phenoxy-thiols:
  • [(HOBut)(4MP)3M(μ-4MP)]2, where M = Ti, Zr, Hf
  • [(py)2M(4MP)], where M = Ti, Zr; py = pyridine
  • [(py)(4MP)3Hf(μ-4MP)]2
4-Mercaptophenol may be used in the synthesis of the following:
  • poly(ethersulfide)s via silylation followed by polycondensation with 2,6-dichloropyridine or 3,6-dichloropyridazine
  • 2,6-di-tertiarybutyl-4-mercaptophenol via Friedel-Craft′s alkylation with tert-butyl chloride in presence of a lewis acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kricheldorf.R.H
Silicon in Polymer Synthesis (2012)
Sriram.D and Yogeeswari.P
Medicinal Chemistry null
Optimization of Ag-Coated Polystyrene Nanosphere Substrates for Quantitative Surface-Enhanced Raman Spectroscopy Analysis.
Ingram WM, et al.
The Journal of Physical Chemistry C, 119(49), 27639-27648 (2015)
New polymer synthesis 99. Hyperbranched poly (ester-imide) s derived from 4, 5-dichlorophthalic acid.
Kricheldorf HR, et al.
High Performance Polymers, 10(3), 217-229 (1998)
Nouf N Mahmoud et al.
Biomaterials science, 8(6), 1669-1682 (2020-01-28)
The blood brain barrier (BBB) is a very selective barrier that protects the brain and the central nervous system (CNS) from the entry of harmful substances and helps regulate the exchange of different molecules and nutrients from and into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service