Skip to Content
Merck
All Photos(1)

Key Documents

541672

Sigma-Aldrich

4-(tert-Butyldimethylsilyloxy)-1-butyne

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH≡C(CH2)2OSi(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
184.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.43 (lit.)

bp

70-74 °C/10 mmHg (lit.)

density

0.893 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)OCCC#C

InChI

1S/C10H20OSi/c1-7-8-9-11-12(5,6)10(2,3)4/h1H,8-9H2,2-6H3

InChI key

NUZBJLXXTAOBPH-UHFFFAOYSA-N

Application

4-(tert-Butyldimethylsilyloxy)-1-butyne may be used in the synthesis of 1-(tert-butyldimethylsilyloxy)-3-pentyn-5-ol and 1-(tert-butyldimethylsilyloxy)-6,6-dimethyl-3-heptyn-5-ol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.9 °F - closed cup

Flash Point(C)

58.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dihydrofurans via an Intramolecular Alkoxide Addition to an Allenylphosphine Oxide.
Pravia K, et al.
The Journal of Organic Chemistry, 61(17), 6031-6032 (1996)

Articles

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Alkynes' versatility enables reactions like addition, metathesis, hydroboration, cleavage, coupling, and cycloadditions in synthetic chemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service