Skip to Content
Merck
All Photos(1)

Key Documents

534099

Sigma-Aldrich

2-Bromophenethylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
200.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5770 (lit.)

bp

253 °C (lit.)

density

1.396 g/mL at 25 °C (lit.)

SMILES string

NCCc1ccccc1Br

InChI

1S/C8H10BrN/c9-8-4-2-1-3-7(8)5-6-10/h1-4H,5-6,10H2

InChI key

ITRNQMJXZUWZQL-UHFFFAOYSA-N

General description

May darken in storage

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel B Straus et al.
ACS nano, 14(3), 3621-3629 (2020-03-03)
We report a family of two-dimensional hybrid perovskites (2DHPs) based on phenethylammonium lead iodide ((PEA)2PbI4) that show complex structure in their low-temperature excitonic absorption and photoluminescence (PL) spectra as well as hot exciton PL. We replace the 2-position (ortho) H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service