Skip to Content
Merck
All Photos(1)

Key Documents

523062

Sigma-Aldrich

3-Vinylbenzaldehyde

97%

Synonym(s):

3-Ethenylbenzaldehyde, 3-Formylstyrene, m-Formylstyrene, m-Vinylbenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4CHO
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.58 (lit.)

density

1.04 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1cccc(C=C)c1

InChI

1S/C9H8O/c1-2-8-4-3-5-9(6-8)7-10/h2-7H,1H2

InChI key

CATOVPRCMWIZLR-UHFFFAOYSA-N

General description

3-Vinylbenzaldehyde (3-VBAL) contains a vinyl group bonded to the aromatic ring of benzaldehyde. Pulsed plasma polymerization of 3-VBAL has been described.

Application

3-Vinylbenzaldehyde may be used in the preparation of:
  • 2-(3-vinylphenyl)-1,3-dioxolane
  • various substituted imines
  • monomer bearing a 2,4,5-triphenylimidazole moiety
  • poly(3-vinylbenzaldehyde-co-dimethylacrylamide) (PVBA-co-PDMA) copolymers

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New benzylidene oxazolone derived polymeric photoswitches for light-induced tunable thermoresponsive behaviors.
Balamurugan A and Lee H.
Polym. Chem., 5(22), 6426-6430 (2014)
A novel de-cross-linking system from cross-linked polymer to linear polymer utilizing pressure or visible light irradiation.
Iwamura T and Sakaguchi M.
Macromolecules, 41(23), 8995-8999 (2008)
R P Garrod et al.
Langmuir : the ACS journal of surfaces and colloids, 23(2), 689-693 (2007-01-11)
A simple two-step plasmachemical methodology is outlined for the fabrication of microcondensor surfaces. This comprises the creation of a superhydrophobic background followed by pulsed plasma deposition of a hydrophilic polymer array. Microcondensation efficiency has been explored in terms of the
Protection and polymerization of functional monomers. 15. Anionic living polymerizations of 2-(3-vinylphenyl)-1, 3-dioxolane and related monomers.
Ishizone T, et al.
Macromolecules, 24(7), 1449-1454 (1991)
Laura L Santos et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(33), 8196-8203 (2009-07-18)
Substituted imines, alpha,beta-unsaturated imines, substituted secondary amines, and beta-amino carbonyl compounds have been synthesized by means of new cascade reactions with mono- or bifunctional gold-based solid catalysts under mild reaction conditions. The related synthetic route involves the hydrogenation of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service