Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Available to ship TODAYfromMILWAUKEE | $143.00 $107.25 |
About This Item
Linear Formula:
(CH3)2NC6H4COCF3
CAS Number:
Molecular Weight:
217.19
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
97%
$107.25
List Price$143.00Save 25%Web-Only Promotion
Available to ship TODAYDetails
Subscriptions are currently unavailable.
Ships Every 4 weeks
Quality Segment
assay
97%
mp
72-76 °C (lit.)
functional group
amine, fluoro, ketone
SMILES string
CN(C)c1ccc(cc1)C(=O)C(F)(F)F
InChI
1S/C10H10F3NO/c1-14(2)8-5-3-7(4-6-8)9(15)10(11,12)13/h3-6H,1-2H3
InChI key
NDKSWWUQHXZADC-UHFFFAOYSA-N
1 of 1
This Item | |||
|---|---|---|---|
| assay 97% | assay 97% | assay 97% | assay 97% |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| mp 72-76 °C (lit.) | mp 19-20 °C (lit.) | mp 151-155 °C (lit.) | mp 31-35 °C (lit.) |
| functional group amine, ketone, fluoro | functional group fluoro, ketone | functional group ketone | functional group bromo, fluoro, ketone |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




