483672
2,3-Dimethyl-1,3-butadiene-d10
≥98 atom % D, ≥99% (CP), contains hydroquinone as stabilizer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
isotopic purity
≥98 atom % D
Assay
≥99% (CP)
contains
hydroquinone as stabilizer
refractive index
n20/D 1.436 (lit.)
bp
68-69 °C (lit.)
mp
−76 °C (lit.)
density
0.815 g/mL at 25 °C (lit.)
mass shift
M+10
SMILES string
[2H]\C([2H])=C(\C(=C(\[2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C6H10/c1-5(2)6(3)4/h1,3H2,2,4H3/i1D2,2D3,3D2,4D3
InChI key
SDJHPPZKZZWAKF-ZRKGKEHVSA-N
Related Categories
Packaging
Packaged in ampules
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service