Skip to Content
Merck
All Photos(1)

Documents

433594

Sigma-Aldrich

2-Piperidineethanol

technical grade, 90%

Synonym(s):

2-(2-Hydroxyethyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

bp

234 °C (lit.)

mp

38-40 °C (lit.)

SMILES string

OCCC1CCCCN1

InChI

1S/C7H15NO/c9-6-4-7-3-1-2-5-8-7/h7-9H,1-6H2

InChI key

PTHDBHDZSMGHKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for syntehsis of:
Spiroimidazolidinone NPC1L1 inhibitors
P2Y12 antagonists for inhibition of platelet aggregation
Cyclin-dependent kinase inhibitors
Anti-HIV agents
Tetraponerine analogs

Reactant for cyclization of secondary homoallylic amines

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Heat capacity of alkanolamine aqueous solutions.
Chiu L-F and Li M-H.
Journal of Chemical and Engineering Data, 44(6), 1396-1401 (1999)
Kinetics of carbon dioxide reaction with sterically hindered 2-piperidineethanol aqueous solutions.
Shen KP, et al.
Industrial & Engineering Chemistry Research, 30(8), 1811-1813 (1991)
Physicochemical properties of 2-piperidineethanol and its aqueous solutions.
Xu S, et al.
Journal of Chemical and Engineering Data, 37(4), 407-411 (1992)
Absorption of carbon dioxide into aqueous solutions of 2-piperidineethanol: Kinetics analysis.
Paul S, et al.
Industrial & Engineering Chemistry Research, 48(3), 1414-1419 (2008)
Investigations of primary and secondary amine carbamate stability by 1 H NMR spectroscopy for post combustion capture of carbon dioxide.
Fernandes D, et al.
The Journal of Chemical Thermodynamics, 54, 183-191 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service