All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H11BrN2
CAS Number:
Molecular Weight:
203.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
≥97.0%
impurities
≤1.0% water
refractive index
n20/D 1.578
SMILES string
[Br-].C[n+]1ccn(CC=C)c1
InChI
1S/C7H11N2.BrH/c1-3-4-9-6-5-8(2)7-9;/h3,5-7H,1,4H2,2H3;1H/q+1;/p-1
InChI key
KLFDZFIZKMEUGI-UHFFFAOYSA-M
Application
1-Allyl-3-methylimidazolium bromide can be used to prepare:
- A weak gel of chitin.
- Composite gels of chitin/cellulose.
- Methylimidazolium based ionic liquids with varying anions as electrolytes in graphene nanosheet-based supercapacitors.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Preparation of chitin/cellulose composite gels and films with ionic liquids
Takegawa A, et al.
Carbohydrate Polymers, 79(1), 85-90 (2010)
1-Allyl-3-methylimidazolium-based ionic liquids employed as suitable electrolytes for high energy density supercapacitors based on graphene nanosheets electrodes
Zarrougui R, et al.
Journal of Molecular Liquids, 249, 795-804 (2018)
Kamalesh Prasad et al.
International journal of biological macromolecules, 45(3), 221-225 (2009-05-27)
This paper reports the formation of weak gel of chitin with an ionic liquid, 1-allyl-3-methylimidazolium bromide (IL). When a mixture of 5% (w/w) chitin with IL was heated at 100 degrees C for 48h, the clear liquid was obtained. The
Zhi Wang et al.
Dalton transactions (Cambridge, England : 2003), 48(18), 6198-6204 (2019-04-13)
A series of hypergolic ionic liquids (HILs) based on the cyano (1H-1,2,3-triazole-1-yl) dihydroborate anion were synthesized by introducing the {BH2-CN} moiety into the 1,2,3-triazole anion. This introduction allowed us to improve the self-ignition property and decrease the viscosity of ionic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service