Skip to Content
Merck
All Photos(1)

Documents

413143

Sigma-Aldrich

2,2′-Methylenebis(6-tert-butyl-4-ethylphenol)

Synonym(s):

2,2′-Methylenebis[6-(1,1-dimethylethyl)-4-ethylphenol], 2,2-Methylenebis(4-ethyl-6-tert -butylphenol), Bis(3-tert -butyl-5-ethyl-2-hydroxyphenyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H2[C(CH3)3](C2H5)OH]2
CAS Number:
Molecular Weight:
368.55
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

119-122 °C (lit.)

SMILES string

CCc1cc(Cc2cc(CC)cc(c2O)C(C)(C)C)c(O)c(c1)C(C)(C)C

InChI

1S/C25H36O2/c1-9-16-11-18(22(26)20(13-16)24(3,4)5)15-19-12-17(10-2)14-21(23(19)27)25(6,7)8/h11-14,26-27H,9-10,15H2,1-8H3

InChI key

GPNYZBKIGXGYNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Antioxidant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Distribution of 2,2'-methylenebis (4-ethyl-6-tert-butylphenol) in rat tissues].
A Takagi et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (105)(105), 26-28 (1987-01-01)
A Takagi et al.
Journal of applied toxicology : JAT, 16(1), 15-23 (1996-01-01)
Groups of 30 Wistar rats of each sex were treated with 2,2'-methylenebis(4-ethyl-6-tert-butylphenol) (MBEBP) in the diet at levels of 0, 0.03, 0.1 and 0.3% for up to 18 months. In both sexes, survival rates of treated groups were similar to
A Takagi et al.
The Journal of toxicological sciences, 17(3), 135-153 (1992-08-01)
The acute and subchronic toxicity studies on 2,2'-methylenebis (4-ethyl-6-tert-butylphenol) (MBEBP) were conducted using male and female Wistar rats. In acute toxicity test, the LD50 values were estimated to be greater than 10 g/kg BW by oral and intraperitoneal administration in
S Tanaka et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (107)(107), 51-55 (1989-01-01)
2,2'-Methylenebis (4-ethyl-6-tert-buthylphenol), an antioxidant, was given orally to pregnant Wistar rats by stomach intubation at the dose levels of 187, 375 or 750 mg/kg body weight during days 7 to 17 of gestation, and the effects of the compound on
[Distribution of 2,2'-methylenebis (4-ethyl-6-tert-butylphenol) in rat fat].
A Takagi
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (106)(106), 105-106 (1988-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service