Skip to Content
Merck
All Photos(1)

Key Documents

411302

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-propanol-OD

98 atom % D

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-propan(ol-d)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CHOD
CAS Number:
Molecular Weight:
169.04
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

98 atom % D

Assay

99% (CP)

technique(s)

NMR: suitable

refractive index

n20/D 1.3 (lit.)

bp

59 °C (lit.)

mp

−4 °C (lit.)

density

1.605 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC(C(F)(F)F)C(F)(F)F

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H/i10D

InChI key

BYEAHWXPCBROCE-MMIHMFRQSA-N

General description

1,1,1,3,3,3-Hexafluoro-2-propanol-OD(HFIP-OD) is the deuterated form of 1,1,1,3,3,3-Hexafluoro-2-propanol. It isused as an NMR solvent in organic chemistry and biochemistry for studying smallmolecules.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service