Skip to Content
Merck
All Photos(1)

Key Documents

407348

Sigma-Aldrich

Poly(ethylene glycol) phenyl ether acrylate

average MN 324, acrylate, phenoxy, 150-350 ppm MEHQ as inhibitor

Synonym(s):

Ethoxylated phenyl acrylate, Phenol ethoxylate acrylate, Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2CHCO2(CH2CH2O)nC6H5
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

product name

Poly(ethylene glycol) phenyl ether acrylate, average Mn 324, contains 150-350 ppm MEHQ as inhibitor

form

viscous liquid

mol wt

average Mn 324

contains

150-350 ppm MEHQ as inhibitor

refractive index

n20/D 1.502

bp

134 °C/1 mmHg (lit.)

density

1.127 g/mL at 25 °C

Ω-end

phenoxy

α-end

acrylate

SMILES string

C=CC(=O)OCCOCCOc1ccccc1

InChI

1S/C11H12O3/c1-2-11(12)14-9-8-13-10-6-4-3-5-7-10/h2-7H,1,8-9H2

InChI key

RZVINYQDSSQUKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly (ethylene glycol) phenyl ether acrylate is hetero-bi functionalized. MEHQ inhibitor prevents auto polymerization.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Monomers for ophthalmic use aim for purity, reliability, and comfort, driving innovation for affordable contact lenses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service