Skip to Content
Merck
All Photos(1)

Key Documents

406600

Sigma-Aldrich

2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decanediol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2(CF2)8CH2OH
CAS Number:
Molecular Weight:
462.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

132 °C/4 mmHg (lit.)

mp

135-137 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO

InChI

1S/C10H6F16O2/c11-3(12,1-27)5(15,16)7(19,20)9(23,24)10(25,26)8(21,22)6(17,18)4(13,14)2-28/h27-28H,1-2H2

InChI key

NSKCTPBWPZPFHW-UHFFFAOYSA-N

General description

2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decanediol (1H,1H,10H,10H-Perfluorodecane-1,10-diol) is a highly fluorinated decanediol. The molecular orientation and multilayer formation of 1H,1H,10H,10H-perfluorodecane-1,10-diol (FC10diol) at the hexane/water interface has been investigated. The interfacial tension of the hexane solution of 1H,1H,10H,10H-perfluorodecane-1,10-diol (FC10diol) against water has been measured. Its freezing point and density have been determined.

Application

2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decanediol may be used in the preparation of diverse bromo(ester) (macro)initiators, required for the preparation of fluorinated pentablock as well as amphiphilic triblock copolymers with a central polyether segment.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel fluorinated block copolymer architectures fuelled by atom transfer radical polymerization.
Jankova K and Hvilsted S.
Journal of Fluorine Chemistry, 126(2), 241-250 (2005)
Takanori Takiue et al.
The journal of physical chemistry. B, 113(44), 14667-14673 (2009-10-13)
The adsorption of 1H,1H,10H,10H-perfluorodecane-1,10-diol (FC10diol) at the hexane solution/water interface was investigated by the measurement of interfacial tension gamma as a function of pressure p and concentration m1 and the thermodynamic data analysis. The results obtained were compared with those
Molecular orientation and miscibility of fluorinated α,ω-alkanediol and alcohol at the hexane/water interface.
Murakami D, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 354(1), 205-209 (2010)
Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 271-271 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service