Skip to Content
Merck
All Photos(1)

Key Documents

368032

Sigma-Aldrich

2-Fluorophenethylamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH2NH2
CAS Number:
Molecular Weight:
139.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.51 (lit.)

bp

64 °C/0.6 mmHg (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

NCCc1ccccc1F

InChI

1S/C8H10FN/c9-8-4-2-1-3-7(8)5-6-10/h1-4H,5-6,10H2

InChI key

RIKUOLJPJNVTEP-UHFFFAOYSA-N

General description

2-Fluorophenethylamine is an amine.

Application

2-Fluorophenethylamine may be used in the synthesis of ethyl 2-cyano-3-(N-2-fluorophenethylamino)-3-methythioacrylate. It may be used for the derivatization of methoxybenzaldehyde polystyrene resin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and bioactivity of Ethyl 2-Cyano-3-methylthio-3-(N-2-Fluorophenethylamino) Acrylates.
ZHOU J-K, et al.
Journal of Guizhou Educational Institute, 017-017 (2007)
S L Dax et al.
Current medicinal chemistry, 6(3), 255-270 (1999-04-29)
Solid-phase organic synthesis, particularly when used in conjunction with combinatorial techniques, is emerging as a revolutionary technology in chemistry. Multi-component reaction systems are particularly valued because several elements of diversity can be introduced in a single transformation thereby expanding the
Daniel B Straus et al.
ACS nano, 14(3), 3621-3629 (2020-03-03)
We report a family of two-dimensional hybrid perovskites (2DHPs) based on phenethylammonium lead iodide ((PEA)2PbI4) that show complex structure in their low-temperature excitonic absorption and photoluminescence (PL) spectra as well as hot exciton PL. We replace the 2-position (ortho) H

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service