Skip to Content
Merck
All Photos(2)

Key Documents

363693

Sigma-Aldrich

4-tert-Butylcalix[6]arene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H84O6
CAS Number:
Molecular Weight:
973.37
Beilstein:
2611918
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

impurities

<5% 4-tert-butylcalix[8]arene

mp

≥300 °C (lit.)

SMILES string

CC(C)(C)c1cc2Cc3cc(cc(Cc4cc(cc(Cc5cc(cc(Cc6cc(cc(Cc7cc(cc(Cc(c1)c2O)c7O)C(C)(C)C)c6O)C(C)(C)C)c5O)C(C)(C)C)c4O)C(C)(C)C)c3O)C(C)(C)C

InChI

1S/C66H84O6/c1-61(2,3)49-25-37-19-39-27-50(62(4,5)6)29-41(56(39)68)21-43-31-52(64(10,11)12)33-45(58(43)70)23-47-35-54(66(16,17)18)36-48(60(47)72)24-46-34-53(65(13,14)15)32-44(59(46)71)22-42-30-51(63(7,8)9)28-40(57(42)69)20-38(26-49)55(37)67/h25-36,67-72H,19-24H2,1-18H3

InChI key

UOEYZAXKBKAKRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for further derivatization, including halogenation, acylation, and diazo coupling.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 28, 3-3 (1995)
Gutsche, C.D.
Calixarenes (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service