Skip to Content
Merck
All Photos(1)

Key Documents

359513

Sigma-Aldrich

1-Acetylpiperazine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O
CAS Number:
Molecular Weight:
128.17
Beilstein:
112220
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

31-34 °C (lit.)

solubility

methanol: soluble 1 g/10 mL, clear, colorless to faintly greenish-yellow

SMILES string

CC(=O)N1CCNCC1

InChI

1S/C6H12N2O/c1-6(9)8-4-2-7-3-5-8/h7H,2-5H2,1H3

InChI key

PKDPUENCROCRCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Infrared and Raman spectra of 1-acetylpiperazine have been recorded in the region of 4000-40cm-1.

Application

1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nesrin Emir et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 127, 388-395 (2014-03-19)
Infrared and Raman spectra of 1-acetylpiperazine (1-ap) have been recorded in the region of 4000-40cm(-1). The conformational isomers, optimized geometric parameters, normal mode frequencies and corresponding vibrational assignments of 1-ap (C6H12N2O) have been examined by density functional theory (DFT), with
Zu-Ping Wu et al.
Archiv der Pharmazie, 345(3), 175-184 (2011-10-13)
A series of novel 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives were synthesized. The cholinesterase inhibition assays indicated that most synthesized compounds exhibited good activity for acetylcholinesterase (AChE) and high selectivity index of AChE over butyrylcholinesterase (BuChE). Compound 12b exhibited the most potent
Tarek Mohamed et al.
Bioorganic & medicinal chemistry letters, 21(19), 5881-5887 (2011-08-30)
A group of 2-substituted N-(naphth-1-ylmethyl)pyrimidin-4-amines (6a-k) and N-benzhydrylpyrimidin-4-amines (7a-k) in conjunction with varying steric and electronic properties at the C-2 position were designed, synthesized and evaluated as dual cholinesterase and amyloid-β (Aβ)-aggregation inhibitors. The naphth-1-ylmethyl compound 6f (2-(4-cyclohexylpiperazin-1-yl)-N-(naphth-1-ylmethyl)pyrimidin-4-amine) exhibited optimum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service