Skip to Content
Merck
All Photos(1)

Documents

357650

Sigma-Aldrich

4-Chlorodiphenyl ether

99%

Synonym(s):

4-Chlorophenyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OC6H5
CAS Number:
Molecular Weight:
204.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.587 (lit.)

bp

161-162 °C/19 mmHg (lit.)

density

1.193 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(Oc2ccccc2)cc1

InChI

1S/C12H9ClO/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9H

InChI key

PGPNJCAMHOJTEF-UHFFFAOYSA-N

General description

4-Chlorodiphenyl ether is halogenated diphenyl ether. It is an estrogenic contaminant and its effect on the growth of endometrial implants has been investigated. Dehalogenations of 4-chlorodiphenyl ether at 375°C over CaO/Ca(OH)2 has been reported.

Application

4-Chlorodiphenyl ether can be used in the preparation of 4′-chloro-2,2′,3,3′,4,5,5′,6,6′-nonabromodiphenyl ether (Cl-BDE-208), an internal standard used in the analysis of highly brominated diphenyl ethers.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Christiansson et al.
Chemosphere, 63(4), 562-569 (2005-11-22)
Polybrominated diphenyl ethers (PBDEs) have been used extensively as brominated flame retardants (BFRs) in textiles, upholstery and electronics. They are ubiquitous contaminants in wildlife and humans. A low concentration of nonabrominated diphenyl ethers (nonaBDEs) is present in commercial DecaBDE and
Yang C and Pittman JR CU
Hazardous Waste and Hazardous Materials, 13(4), 445-464 (1996)
J Z Yang et al.
Reproductive toxicology (Elmsford, N.Y.), 11(1), 69-75 (1997-01-01)
The objectives of this study were to evaluate the effects of repeated injection of 4-chlorodiphenyl ether (4-CDE) on the survival and growth of endometriotic implants. Endometriosis was surgically induced in adult female B6C3F1 mice (n = 25) after ovariectomy. Seven
Y C Chui et al.
Drug metabolism and disposition: the biological fate of chemicals, 15(1), 44-50 (1987-01-01)
The metabolic disposition of 14C-labeled 4-chlorodiphenyl ether ([14C]4-CDE) was examined in rats following iv administration of a single dose (850 nmol/kg). [14C]4-CDE decayed rapidly from the blood since no unchanged [14C]4-CDE was detected in the blood beyond 2 hr after
W G Foster et al.
Canadian journal of physiology and pharmacology, 75(10-11), 1188-1196 (1998-02-12)
Surgically induced endometriosis in the mouse has been described as a model to investigate the effect of environmental pollutants on the growth of endometriotic implants. The objectives of this study were to evaluate a modified surgical procedure to induce endometriosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service