Skip to Content
Merck
All Photos(1)

Key Documents

337285

Sigma-Aldrich

2,3,4-Trifluorophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2OH
CAS Number:
Molecular Weight:
148.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.465 (lit.)

bp

69 °C/43 mmHg (lit.)

mp

30-34 °C (lit.)

density

1.46 g/mL at 25 °C (lit.)

SMILES string

Oc1ccc(F)c(F)c1F

InChI

1S/C6H3F3O/c7-3-1-2-4(10)6(9)5(3)8/h1-2,10H

InChI key

IJGSULQFKYOYEU-UHFFFAOYSA-N

General description

2,3,4-Trifluorophenol is an ortho-fluorinated trifluorophenol. Biodegradation and oxidative biodehalogenation of 2,3,4-trifluorophenol has been studied by 19F nuclear magnetic resonance (NMR) spectra.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Boersma et al.
Applied and environmental microbiology, 64(4), 1256-1263 (1998-04-18)
A method was developed to study the biodegradation and oxidative biodehalogenation of fluorinated phenols by 19F nuclear magnetic resonance (NMR). Characterization of the 19F NMR spectra of metabolite profiles of a series of fluorophenols, converted by purified phenol hydroxylase, catechol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service