Skip to Content
Merck
All Photos(2)

Key Documents

331317

Sigma-Aldrich

5-Bromovaleryl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)4COCl
CAS Number:
Molecular Weight:
199.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

116-118 °C/33 mmHg (lit.)

density

1.49 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCBr

InChI

1S/C5H8BrClO/c6-4-2-1-3-5(7)8/h1-4H2

InChI key

OKRUMSWHDWKGHA-UHFFFAOYSA-N

Application

5-Bromovaleryl chloride was used in the preparation of aromatic azides and organic gelators.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of the neutral and cationic gelators from (1S, 2S)-(-)-diphenylethylenediamine for the preparation of silica nano tubes.
Hyun MH, et al.
Bull. Korean Chem. Soc., 30(7), 1641-1643 (2009)
Rajavel Srinivasan et al.
Organic letters, 8(4), 713-716 (2006-02-14)
[reaction: see text] We have successfully designed and synthesized a small library of protein tyrosine phosphatase (PTP) inhibitors, in which the so-called "click chemistry" or Cu(I)-catalyzed 1,3-dipolar alkyne-azide coupling reaction was carried out for rapid assembly of 66 different bidentate
Duy Khuong Mai et al.
Molecules (Basel, Switzerland), 25(15) (2020-07-29)
The synthesis of three water-soluble lactose-modified 4,4-difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY)-based photosensitizers with tumor-targeting capabilities is reported, including an investigation into their photodynamic therapeutic activity on three distinct cancer cell lines (human hepatoma Huh7, cervical cancer HeLa, and breast cancer MCF-7 cell lines).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service