Skip to Content
Merck
All Photos(1)

Key Documents

320358

Sigma-Aldrich

Isobutyraldehyde

98%

Synonym(s):

2-Methylpropanal, 2-Methylpropionaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCHO
CAS Number:
Molecular Weight:
72.11
Beilstein:
605330
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.5 (vs air)

vapor pressure

66 mmHg ( 4.4 °C)

Assay

98%

form

liquid

autoignition temp.

384 °F

expl. lim.

10 %, 25 °F
2 %, 32 °F

refractive index

n20/D 1.374 (lit.)

bp

63 °C (lit.)

mp

−65 °C (lit.)

solubility

60 g/L at 25 °C

density

0.79 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)C(C)C

InChI

1S/C4H8O/c1-4(2)3-5/h3-4H,1-2H3

InChI key

AMIMRNSIRUDHCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-11.2 °F - closed cup

Flash Point(C)

-24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rudy Coquet et al.
Physical chemistry chemical physics : PCCP, 9(45), 6040-6046 (2007-11-16)
The formation and catalytic behavior of active structures in a SiO(2)-supported unsaturated Ru complex catalyst for alkene epoxidation were studied by means of hybrid density functional theory (DFT) calculations. An energy-gaining route for the catalyst activation was found to allow
Hamid Reza Khavasi et al.
Inorganic chemistry, 48(13), 5593-5595 (2009-05-28)
Dioxygen epoxidation of cyclic alkenes into their corresponding epoxides was successfully achieved in good yield by using a novel binuclear manganese carboxamide complex as the catalyst and isobutyraldehyde as the cosubstrate.
Organocatalytic enantioselective cross-aldol reactions of aldehydes with isatins: formation of two contiguous quaternary centered 3-substituted 3-hydroxyindol-2-ones.
Fei Xue et al.
Chemistry, an Asian journal, 4(11), 1664-1667 (2009-09-12)
Christine Counet et al.
Journal of agricultural and food chemistry, 50(8), 2385-2391 (2002-04-04)
After vacuum distillation and liquid-liquid extraction, the volatile fractions of dark chocolates were analyzed by gas chromatography-olfactometry and gas chromatography-mass spectrometry. Aroma extract dilution analysis revealed the presence of 33 potent odorants in the neutral/basic fraction. Three of these had
Hyunwoo Kim et al.
Organic letters, 11(1), 157-160 (2008-12-10)
Addition of isobutyraldehyde to 1,2-bis(2-hydroxyphenyl)-1,2-diaminoethane (mother diamine) cleanly gives a stable, fused imidazolidine-dihydro-1,3-oxazine ring complex. However, vigorous heating of the fused ring complex gives the diaza-Cope rearrangement product with excellent yield and stereoselectivity. A variety of alkyl aldehydes have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service