Skip to Content
Merck
All Photos(3)

Documents

311324

Sigma-Aldrich

Platinum on alumina

greener alternative

extent of labeling: 5 wt. % loading, -325 mesh, powder

Synonym(s):

Pt/Al2O3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: platinum
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

surface area

>250 m2/g

particle size

-325 mesh

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Platinum on alumina (Pt/Al2O3) (5 wt. % loading, -325 mesh, powder) can be used as a catalyst for a variety of applications, such as:
  • in the hydrodechlorination of chlorobenzenes
  • oxidation, reduction, and decomposition
  • in methanol steam reforming
  • hydrocarbon combustion

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paquette, L. A.; Miyahara, Y.; Doecke, C. W.
Journal of the American Chemical Society, 108, 1716-1716 (1986)
Olah, G. A.; Gupta, B. et al.
Journal of the American Chemical Society, 107, 7097-7097 (1985)
Fraenkel, G.; Gallucci, J.; Rosenzweig, H. S.
The Journal of Organic Chemistry, 54, 677-677 (1989)
Kinetics of o-Xylene Combustion over a Pt/Alumina Catalyst
Manta CM, et al.
Chemical Engineering & Technology, 35(12), 2147-2154 (2012)
Preparation of Platinum-Supported Alumina Lewis Superacid Catalyst Using a CVD Method
Ayame A, et al.
Studies in Surface Science and Catalysis, 90, 257-262 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service