Skip to Content
Merck
All Photos(2)

Key Documents

292710

Sigma-Aldrich

Phenyl β-D-glucopyranoside

≥95.0%

Synonym(s):

Phenyl beta-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O6
CAS Number:
Molecular Weight:
256.25
Beilstein:
87517
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

powder

optical activity

[α]25/D −70°, c = 1 in H2O

mp

176-178 °C (lit.)

SMILES string

OC[C@H]1O[C@@H](Oc2ccccc2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H16O6/c13-6-8-9(14)10(15)11(16)12(18-8)17-7-4-2-1-3-5-7/h1-5,8-16H,6H2/t8-,9-,10+,11-,12-/m1/s1

InChI key

NEZJDVYDSZTRFS-RMPHRYRLSA-N

Application

Phenyl β-D-glucopyranoside can be used:
  • As a starting material for the synthesis of various derivatives of β-D-glucopyranosides with potential application as anti-HIV agents.
  • As a model for glycosides in the gas phase for their spectroscopic investigation.
  • As an internal standard in GC and GC-MS quantitative analyses.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4-(Arylamino)phenyl alpha-D-glucopyranosides as potential anti-HIV agents.
J C Briggs et al.
Carbohydrate research, 282(2), 293-298 (1996-03-18)
Sugars in the gas phase: the spectroscopy and structure of jet-cooled phenyl ?-D-glucopyranoside.
Talbot FO and Simons JP
Physical Chemistry Chemical Physics, 4(15), 3562-3565 (2002)
Chunyan Bao et al.
Carbohydrate research, 339(7), 1311-1316 (2004-04-29)
A new hydrogel based on a substituted phenyl glucoside with a Schiff base in the aglycon was synthesized, and the self-assembling characteristics was studied. FTIR spectra, UV-vis absorption spectra and X-ray diffraction (XRD) revealed that pi-pi interactions between the Schiff
Ying Na et al.
Bioorganic chemistry, 39(3), 111-113 (2011-03-26)
The spontaneous hydrolysis of glycosylamines, where the aglycone is either a primary amine or ammonia, is over a hundred million-times faster than that of O- or S-glycosides. The reason for this (as pointed out by Capon and Connett in 1965)
Emission of 2-phenylethanol from its ?-D-glucopyranoside and the biogenesis of these compounds from [2H8] L-phenylalanine in rose flowers.
Hayashi S, et al.
Tetrahedron, 60(33), 7005-7013 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service