Skip to Content
Merck
All Photos(1)

Key Documents

272310

Sigma-Aldrich

5-Methylquinoxaline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.62 (lit.)

bp

120 °C/15 mmHg (lit.)

mp

20-21 °C (lit.)

density

1.125 g/mL at 25 °C

SMILES string

Cc1cccc2nccnc12

InChI

1S/C9H8N2/c1-7-3-2-4-8-9(7)11-6-5-10-8/h2-6H,1H3

InChI key

CQLOYHZZZCWHSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Methylquinoxaline was used in the synthesis of 1,12-diphenyl-5-methyl-1,12,12a,12b-tetrahydrobis[1,2,4]triazolo[4,3-a:3′,4′-c]quinoxaline bis-cycloadducts.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactivity of asymmetric benzo-condensed diazines with nitrilimine dipoles in the 1, 3-dipolar cycloaddition reactions.
Tetrahedron Letters, 50(52), 7333-7336 (2009)
Ecem Berk et al.
Journal of agricultural and food chemistry, 67(17), 4923-4930 (2019-04-11)
This study investigated the formation of Maillard reaction products in sesame seeds under different roasting conditions. Sesame seeds were roasted at 150, 180, 200, and 220 °C for 10 min, and thermal process contaminants including 5-hydroxymethylfurfural, acrylamide, furan, and dicarbonyl
Weerachat Sompong et al.
BMC complementary and alternative medicine, 15, 394-394 (2015-11-02)
Methylglyoxal (MG) is one of the most reactive glycating agents, which result the formation of advanced glycation end-products (AGEs) that have been implicated in the progression of age-related diseases. Inhibition of MG-induced AGE formation is the imperative approach for alleviating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service