Skip to Content
Merck
All Photos(3)

Documents

257710

Sigma-Aldrich

Selenium sulfide

Synonym(s):

Selenium disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SeS2
CAS Number:
Molecular Weight:
143.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

form

powder

SMILES string

S=[Se]=S

InChI

1S/S2Se/c1-3-2

InChI key

JNMWHTHYDQTDQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selenium sulfide (SeS2) can be used in a variety of applications such as:
  • antifungal agent in shampoos.
  • as electrode material in lithium ion batteries.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer A Stein et al.
Pediatric dermatology, 22(4), 331-333 (2005-08-03)
We describe three teenage siblings with confluent and reticulated papillomatosis, all presenting during a 6-month period. Two of the three patients had confirmed tinea versicolor, with positive potassium hydroxide scrapings, in association with this entity. This is the largest series
Selenium sulfide@ mesoporous carbon aerogel composite for rechargeable lithium batteries with good electrochemical performance
Zhang Z, et al.
Journal of Power Sources, 284(12), 95-102 (2015)
Holger Fleischer et al.
Dalton transactions (Cambridge, England : 2003), (19)(19), 3221-3228 (2005-09-21)
The molecular structures of Se(SCH(3))(2) and Te(SCH(3))(2) were investigated using gas-phase electron diffraction (GED) and ab initio and DFT geometry optimisations. While parameters involving H atoms were refined using flexible restraints according to the SARACEN method, parameters that depended only
Noel Prevost et al.
Journal of drugs in dermatology : JDD, 7(7), 689-691 (2008-07-31)
Hair shaft discoloration has been documented to be caused by disease states, medications, and exogenous chemicals. After researching the literature, xanthotrichia or yellow hair has been determined to be caused predominately by exogenous chemicals. Two cases of new chemicals causing
Cheryl A Hull et al.
Cutis, 73(6), 425-429 (2004-07-01)
Pityriasis (tinea) versicolor, which consists of hyperpigmented and hypopigmented scaly patches, is often difficult to treat. A double-blind comparative study between once-a-day sodium sulfacetamide lotion and selenium sulfide lotion was undertaken. Both treatments were safe and efficacious. Selenium sulfide was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service