Skip to Content
Merck
All Photos(1)

Key Documents

254487

Sigma-Aldrich

Di-tert-butyl malonate

98%

Synonym(s):

1,3-Bis(1,1-dimethylethyl) propanedioate, 1,3-Di-tert-butyl propanedioate, Bis(1,1-dimethylethyl) malonate, di-tert-butyl propanedioate, tert-Butyl malonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCOCH2COOC(CH3)3
CAS Number:
Molecular Weight:
216.27
Beilstein:
1781766
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.418 (lit.)

bp

110-111 °C/22 mmHg (lit.)

mp

−7-−6 °C (lit.)

density

0.966 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)OC(=O)CC(=O)OC(C)(C)C

InChI

1S/C11H20O4/c1-10(2,3)14-8(12)7-9(13)15-11(4,5)6/h7H2,1-6H3

InChI key

CLPHAYNBNTVRDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Di-tert-butyl malonate was used in total synthesis of (+/−)-actinophyllic acid. It was also employed as precursor for metal-organic chemical vapor deposition of HfO2 and ZrO2 thin films.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service