Skip to Content
Merck
All Photos(1)

Key Documents

250236

Sigma-Aldrich

1-(tert-Butyldimethylsilyl)imidazole

≥95%

Synonym(s):

TBDMSIM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2Si
CAS Number:
Molecular Weight:
182.34
Beilstein:
606695
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

53 °C/0.2 mmHg (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)n1ccnc1

InChI

1S/C9H18N2Si/c1-9(2,3)12(4,5)11-7-6-10-8-11/h6-8H,1-5H3

InChI key

VUENSYJCBOSTCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(tert-Butyldimethylsilyl)imidazole is a reactive silylating agent.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D C Landrum et al.
Journal of chromatography, 483, 21-32 (1989-12-08)
The use of gas-liquid chromatography and mass spectrometry with derivatizing agents that give stable derivatives and consistent fragmentation patterns allows for accurate identification of a variety of compounds. In this study either N-methyl-N-tert.-butyldimethylsilyltrifluoroacetamide or N-tert.-butyldimethylsilylimidazole were employed to derivatize a
P Bydal et al.
Steroids, 61(6), 349-353 (1996-06-01)
Five dehydrated compounds obtained from a tert-butyldimethylsilylchloride/imidazole or an aqueous hydrochloric acid treatment of 17 alpha-butyl-3-O-methyl estradiol in refluxing solvent were purified and characterized. Three compounds were obtained from a direct vicinal proton elimination, the two others from a vicinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service