Skip to Content
Merck
All Photos(1)

Documents

247898

Sigma-Aldrich

Isosorbide dimethyl ether

98%

Synonym(s):

DMI, Dimethyl isosorbide, 1,4:3,6-Dianhydro-2,5-di-O-methyl-D-glucitol, 1,4:3,6-Dianhydrosorbitol 2,5-dimethyl ether, 2,5-Di-O-methyl-1,4:3,6-dianhydro-D-glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
Beilstein:
80854
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]21/D +96.1°, c = 2 in chloroform

refractive index

n20/D 1.461 (lit.)

bp

93-95 °C/0.1 mmHg (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CO[C@H]1COC2[C@@H](COC12)OC

InChI

1S/C8H14O4/c1-9-5-3-11-8-6(10-2)4-12-7(5)8/h5-8H,3-4H2,1-2H3/t5-,6+,7-,8-/m1/s1

InChI key

MEJYDZQQVZJMPP-ULAWRXDQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isosorbide dimethyl ether (DMI), a sustainable solvent, is widely used in various cosmetic and pharmaceutical formulations.
General applications:
  • DMI can be used as an alternative green solvent for epoxidation of cyclooctene, Baylis–Hillman reaction of isomaltulose-derived aldehydes with α,β-unsaturated ketones and also in solid-phase synthesis.
  • It can be used as a coalescent for water-borne paints, where it acts as a co-solvent during the water loss phase and as a coalescing agent during the film formation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Brisaert et al.
Pharmacy world & science : PWS, 18(5), 182-186 (1996-10-01)
Erythromycin, an antibiotic commonly used topically in the treatment of acne, is unstable in solution. The stability is influenced by the pH and by the presence of water. The influence of the pH on the stability of erythromycin was investigated
Y Zhang et al.
International journal of cosmetic science, 41(5), 479-488 (2019-08-06)
Phenylethyl resorcinol (PR) has been used widely in the personal care industry as a novel skin lightening ingredient. Surprisingly, there is only limited information describing the physicochemical properties of this active. Therefore, the primary objective of this study was to
B W Barry et al.
The Journal of investigative dermatology, 82(1), 49-52 (1984-01-01)
The occluded vasoconstrictor assay was used to assess the effect of penetration enhancers on the topical bioavailability of a representative steroid, betamethasone 17-benzoate, with dimethylisosorbide (DMI) as a reference solvent. In the initial test, only N-methyl-2-pyrrolidone (NMP) increased the steroid
Isosorbide mono-and di-alkyl ethers, a new class of sustainable coalescents for water-borne paints.
Durand M, et al.
Progress in Organic Coatings, 69(4), 344-351 (2010)
A Otto et al.
Skin pharmacology and physiology, 21(6), 326-334 (2008-10-04)
In this study the effect of 2 penetration modifiers, dimethyl isosorbide (DMI) and diethylene glycol monoethyl ether (DGME) on the skin delivery of hydroquinone (HQ), salicylic acid (SA) and octadecenedioic acid (DIOIC) was investigated. Ten percent DMI and DGME were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service