Skip to Content
Merck
All Photos(1)

Key Documents

247049

Sigma-Aldrich

Tributylphosphine

mixture of isomers, 97%

Synonym(s):

P(n-Bu)3, TBP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]3P
CAS Number:
Molecular Weight:
202.32
Beilstein:
1738261
EC Number:
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

vapor density

9 (vs air)

Quality Level

Assay

97%

form

liquid

autoignition temp.

392 °F

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Acetylations

reagent type: ligand
reaction type: Addition Reactions

reagent type: ligand
reaction type: Stille Coupling

impurities

<1% TBP oxide
5% TBP isomers

refractive index

n20/D 1.462 (lit.)

bp

150 °C/50 mmHg (lit.)

density

0.81 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CCCCP(CCCC)CCCC

InChI

1S/C12H27P/c1-4-7-10-13(11-8-5-2)12-9-6-3/h4-12H2,1-3H3

InChI key

TUQOTMZNTHZOKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-addition catalyst; used with disulfides for the thioetherification of alcohols; acylation catalyst; used to prepare active esters.
Used with platinum (II) or (IV) in an intermolecular hydroamination of unactivated alkenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1A

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

242.6 °F - closed cup

Flash Point(C)

117 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Mol. Catal. A: Chem., 271, 145-145 (2007)
J Krijt et al.
Clinical chemistry, 47(10), 1821-1828 (2001-09-25)
Aminothiols have been implicated in the pathogenesis of arteriosclerosis, and reliable methods are needed to determine their concentrations in body fluids. We present a comparison of two analytical methods and focus on the reduction of low-molecular weight and protein-mixed disulfides
J X Yan et al.
Journal of chromatography. A, 813(1), 187-200 (1998-08-11)
A simple technique is introduced to identify and quantitate cysteine (Cys) after acid hydrolysis of protein. The technique involves using 9-fluorenylmethyl chloroformate (Fmoc)-based amino acid analysis that recovers all of the amino acids (asparagine and glutamine are recovered in their
Jing-Yu Wu et al.
The Journal of organic chemistry, 73(22), 9137-9139 (2008-10-24)
Aziridines underwent cyclization reaction with carbon disulfide and isothiocyanate in the presence of organophosphine to afford thiazolidinone derivatives in good to high yields. The mechanistic study revealed that organophosphine serves as a catalyst in the reaction.
Yin-Wei Sun et al.
Organic letters, 12(24), 5664-5667 (2010-11-13)
P(n)Bu(3)-catalyzed cyclization reactions of salicylaldimines and salicylaldehydes with ethyl 2,3-butadienoate gave the corresponding functionalized chromans in moderate to good yields in THF under mild conditions. The new reaction provides a new method for the synthesis of biologically active chroman products.

Articles

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Solvothermal synthesis of nanoparticles: applications from nanocircuits and nano-optical circuits to nanomagnetics and biotech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service