Skip to Content
Merck
All Photos(1)

Documents

232181

Sigma-Aldrich

Methyl 1-cyclohexene-1-carboxylate

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H9CO2CH3
CAS Number:
Molecular Weight:
140.18
Beilstein:
1071971
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

bp

190-192 °C
58-60 °C/0.1 mmHg (lit.)

density

1.028 g/mL at 20 °C (lit.)

SMILES string

COC(=O)C1=CCCCC1

InChI

1S/C8H12O2/c1-10-8(9)7-5-3-2-4-6-7/h5H,2-4,6H2,1H3

InChI key

KXPWRCPEMHIZGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl 1-cyclohexene-1-carboxylate was used in:
  • diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols
  • synthesis of methyl 7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate
  • synthesis of ethyl 7-oxo-7b,8,9,10,11,11a-hexahydro-7H-benzo[b]phenaleno[2,1-d]furan-11a-carboxylate

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diastereoselective Synthesis of cis-1, 2-Dialkenylcyclopropanols and Subsequent Oxy-Cope Rearrangement.
Lee J, et al.
Journal of the American Chemical Society, 117(39), 9919-9920 (1995)
Ceric ammonium nitrate (CAN)-mediated oxidative cycloaddition of 1, 3-dicarbonyls to conjugated compounds. Efficient synthesis of dihydrofurans, dihydrofurocoumarins, dihydrofuroquinolinones, dihydrofurophenalenones, and furonaphthoquinone natural products.
Lee YR, et al.
Tetrahedron, 56(45), 8845-8853 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service