Skip to Content
Merck
All Photos(2)

Key Documents

204579

Sigma-Aldrich

Terbium(III,IV) oxide

99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tb4O7
CAS Number:
Molecular Weight:
747.70
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: terbium

density

7.3 g/mL at 25 °C (lit.)

SMILES string

O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O

InChI

1S/7O.4Tb

InChI key

UWZVBPNIJCKCMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service