Skip to Content
Merck
All Photos(1)

Key Documents

192309

Sigma-Aldrich

3,3-Dimethyloxetane

98%

Synonym(s):

β,β-Dimethyloxetane, 3,3-Dimethyloxacyclobutane, 3,3-Dimethyloxetane, 3,3-Dimethyltrimethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.399 (lit.)

bp

81 °C/765 mmHg (lit.)

density

0.835 g/mL at 25 °C (lit.)

SMILES string

CC1(C)COC1

InChI

1S/C5H10O/c1-5(2)3-6-4-5/h3-4H2,1-2H3

InChI key

RVGLUKRYMXEQAH-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Billing et al.
The Journal of organic chemistry, 77(24), 11227-11231 (2012-11-16)
A novel one-step method for mildly converting cyclic ethers into dibromo compounds is reported. Alcohols, oximes, aldehydes, and ketones are known to react under Appel or Corey-Fuchs reaction conditions, but apparently these have never been applied to oxetanes or larger
Copolymerization of oxetane with 3, 3-dimethyloxetane-I. Reactivity parameters and microstructure of the copolymers.
Bucquoye M and Goethals EJ.
Eur. Polymer J., 14(5), 323-328 (1978)
Cyclic oligomers in the cationic polymerization of 3, 3-dimethyloxetane.
Bucquoye M and Goethals EJ.
Makromol. Chem., 179(7), 1681-1688 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service