Skip to Content
Merck
All Photos(2)

Key Documents

183636

Sigma-Aldrich

2-Iodobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4CH2OH
CAS Number:
Molecular Weight:
234.03
Beilstein:
2079487
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

89-92 °C (lit.)

SMILES string

OCc1ccccc1I

InChI

1S/C7H7IO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2

InChI key

WZCXOBMFBKSSFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Iodobenzyl alcohol was used in the synthesis of:
  • substituted seven-membered lactones
  • 2-[(E)-(1′-iodo-2′-propenyl)]benzyl alcohol
  • 2,3-diphenyl-1-indenone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of indenones via palladium-catalyzed annulation of internal alkynes.
Larock RC, et al.
The Journal of Organic Chemistry, 58(17), 4579-4583 (1993)
Dinesh Kumar Rayabarapu et al.
Journal of the American Chemical Society, 124(20), 5630-5631 (2002-05-16)
A new class of substituted seven-membered lactones 3 were conveniently synthesized via cyclization of o-iodobenzyl alcohol 1 (o-IC(6)H(4)CH(2)OH) with various propiolates 2 (RC triple bond CCOOMe) in the presence of Ni(dppe)Br(2) and Zn powder in acetonitrile at 80 degrees C.
Guangwei Wang et al.
Journal of organometallic chemistry, 692(21), 4731-4736 (2007-10-01)
The Zr-catalyzed methylalumination of heterosubstituted arylethynes containing O, S, Cl, and Si can proceed in high yields (>70%) and in a highly regio- and stereoselective manner (≥98-99%), although SO(2)Ph, Br, and Cl in a benzylic position present serious chemoselectivity-related problems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service