Skip to Content
Merck

175692

Aniline-2,3,4,5,6-d5

98 atom % D

Synonym(s):

2,3,4,5,6-Pentadeuterioaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6D5NH2
CAS Number:
Molecular Weight:
98.16
UNSPSC Code:
12352200
NACRES:
NA.12
PubChem Substance ID:
EC Number:
224-015-9
Beilstein/REAXYS Number:
2208322
MDL number:
Isotopic purity:
98 atom % D
Assay:
99% (CP)
Mass shift:
M+5
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.22 (184 °C, vs air)

Quality Segment

vapor pressure

0.7 mmHg ( 25 °C)

isotopic purity

98 atom % D

assay

99% (CP)

expl. lim.

11 %

refractive index

n20/D 1.5836 (lit.)

bp

184 °C (lit.)

mp

-6 °C (lit.)

density

1.076 g/mL at 25 °C

mass shift

M+5

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c([2H])c(N)c([2H])c1[2H]

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i1D,2D,3D,4D,5D

InChI key

PAYRUJLWNCNPSJ-RALIUCGRSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

target_organs

Blood

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

158.0 °F - closed cup

flash_point_c

70 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library