Skip to Content
Merck
All Photos(2)

Key Documents

153044

Sigma-Aldrich

3-Aminopyrazole-4-carbonitrile

97%

Synonym(s):

3-Amino-4-cyanopyrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N4
CAS Number:
Molecular Weight:
108.10
Beilstein:
2647
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

172-174 °C (lit.)

SMILES string

Nc1n[nH]cc1C#N

InChI

1S/C4H4N4/c5-1-3-2-7-8-4(3)6/h2H,(H3,6,7,8)

InChI key

FFNKBQRKZRMYCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-amino-4-cyanopyrazole was used to study the impurities in zaleplon bulk drug.
Building block used in the synthesis of a variety of heterocycles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ch Bharathi et al.
Journal of pharmaceutical and biomedical analysis, 44(1), 101-109 (2007-03-21)
Zaleplon is a pyrazolopyrimidine derivative and possesses sedative and hypnotic properties. Seven unknown impurities in zaleplon bulk drug at levels below 0.1% were detected by reverse-phase high performance liquid chromatography (HPLC). The starting material, 3-amino-4-cyanopyrazole and an intermediate, N-[3-[3-(dimethylamino)-1-oxo-2-propenyl]-phenyl]-N-ethylacetamide (DOPEA)
Synlett, 65-65 (2006)
Helvetica Chimica Acta, 89, 30-30 (2006)
Alessandro Antonelli et al.
Molecular and cellular endocrinology, 393(1-2), 56-64 (2014-06-17)
CLM29 (a pyrazolo[3,4-d]pyrimidine, that inhibits RET, epidermal growth factor receptor, vascular endothelial growth factor receptor, and has an anti-angiogenic activity) has anti-neoplastic activity in papillary dedifferentiated thyroid cancer. Here we tested CLM29 in medullary thyroid cancer (MTC), in primary MTC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service