Skip to Content
Merck
All Photos(1)

Key Documents

14865

Sigma-Aldrich

3-[Bis(2-hydroxyethyl)amino]propyl-triethoxysilane solution

technical, ~65% in ethanol

Synonym(s):

N-(3-Triethoxysilylpropyl)diethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H31NO5Si
CAS Number:
Molecular Weight:
309.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

form

liquid

concentration

~65% in ethanol

SMILES string

CCO[Si](CCCN(CCO)CCO)(OCC)OCC

InChI

1S/C13H31NO5Si/c1-4-17-20(18-5-2,19-6-3)13-7-8-14(9-11-15)10-12-16/h15-16H,4-13H2,1-3H3

InChI key

IYAYDWLKTPIEDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-[Bis(2-hydroxyethyl)amino]propyl-triethoxysilane solution was used to functionalize nanoporous silica (SBA-15).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.0 °F

Flash Point(C)

23.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Payman Hashemi et al.
Analytica chimica acta, 646(1-2), 1-5 (2009-06-16)
Nanoporous silica (SBA-15) was prepared and functionalized with 3-[Bis(2-hydroxyethyl)amino] propyl-triethoxysilane (HPTES) to be used as a highly porous fiber coating material for solid-phase microextraction (SPME). The prepared HPTES-SBA-15 particles had a lengthy morphology and a specific surface area of 790

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service