Skip to Content
Merck
All Photos(3)

Key Documents

140023

Sigma-Aldrich

Tetraethylammonium bromide

reagent grade, 98%

Synonym(s):

TEAB, TEA bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Br)
CAS Number:
Molecular Weight:
210.16
Beilstein:
3563430
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
grade:
reagent grade
Assay:
98%

grade

reagent grade

Quality Level

Assay

98%

form

crystalline

impurities

≤2% triethylamine hydrobromide

pH

6.5 (100 g/L)

mp

285 °C (dec.) (lit.)

SMILES string

[Br-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BrH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

HWCKGOZZJDHMNC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tetraethylammonium bromide may be used:
  • As an organic template for synthesizing zeolite beta via hydrothermal crystallization.
  • As a catalyst for the oxidative coupling of aldehydes or alcohols with thiophenols or disulfides to form thioesters.
  • In combination with o-iodoxybenzoic acid (IBX) for the oxidative dehomologation of primary carboxamides to nitriles and α,α-disubstituted acetamides to ketones. This reagent combination can also be used for the conversion of sulfides to sulfoxides and N,N-disubstituted glycylamides into corresponding cyanamides.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

361.4 °F - closed cup

Flash Point(C)

183 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel and facile transformation of N, N-disubstituted glycylamides into corresponding cyanamides by using pentavalent iodine reagents in combination with tetraethylammonium bromide.
Chaudhari K
Synlett, 18, 2815-2818 (2007)
Oxidative Conversion of ?, ?-Disubstituted Acetamides to Corresponding One-Carbon-Shorter Ketones Using Hypervalent Iodine (?5) Reagents in Combination with Tetraethylammonium Bromide.
Bellale E
The Journal of Organic Chemistry, 73(23), 9473-9475 (2008)
Hydrothermal crystallization of zeolite beta using tetraethylammonium bromide.
Eapen M
Zeolites, 14(4), 295-302 (1994)
A mild, chemoselective oxidation of sulfides to sulfoxides using o-iodoxybenzoic acid and tetraethylammonium bromide as catalyst.
Shukla V
The Journal of Organic Chemistry, 68(13), 5422-5425 (2003)
o-Iodoxybenzoic acid-and tetraethylammonium bromide-mediated oxidative transformation of primary carboxamides to one-carbon dehomologated nitriles.
Bhalerao D
The Journal of Organic Chemistry, 72(2), 662-665 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service